Learn More
Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents.
£60.23 - £436.27
Specifications
For Use With (Equipment) | Ion Selective Electrode |
---|---|
Type | Ion Selective Electrode Reagents |
For Use With (Application) | Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11722787
|
Thermo Scientific™
841101 |
Storage Solution for Sodium Electrode | 475 mL |
£66.00
Each |
In Stock
Log in to see stock. |
||||
12049967
|
Thermo Scientific™
841111 |
Ionic Strength Adjustor Buffer Solution for Sodium Ion Selective Electrodes, 475mL | 475 mL |
£60.23
475mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12038882
|
Thermo Scientific™
841113 |
Reconditioning Solution for Sodium Electrode | 475 mL |
£98.96
475mL |
In Stock
Log in to see stock. |
||||
11767527
|
Thermo Scientific™
930710 |
Nitrate Interference Suppressor Solution for Nitrate Electrode | 475 mL |
£89.49
475mL |
In Stock
Log in to see stock. |
||||
11777527
|
Thermo Scientific™
930711 |
ISA for Fluoroborate, Nitrate, Perchlorate Electrodes | 475 mL |
£89.49
475mL |
In Stock
Log in to see stock. |
||||
11717537
|
Thermo Scientific™
931911 |
ISA for Potassium Electrode | 475 mL |
£107.56
475mL |
In Stock
Log in to see stock. |
||||
11747537
|
Thermo Scientific™
932011 |
ISA for Calcium Electrode | 475 mL |
£105.84
475mL |
In Stock
Log in to see stock. |
||||
11976079
|
Thermo Scientific™
940011 |
ISA for Bromide, Cadmium, Chloride, Cupric, Iodide, Sufide, Thiocyanate Electrodes | 475 mL |
£86.91
475mL |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
11757647
|
Thermo Scientific™
940909 |
TISAB II for Fluoride Electrode | 1 Gal. |
£140.26
3.80 L |
In Stock
Log in to see stock. |
||||
11966089
|
Thermo Scientific™
940911 |
TISAB III Concentrate for Fluoride Electrode | 475 mL |
£150.59
475mL |
In Stock
Log in to see stock. |
||||
10665261
|
Thermo Scientific™
940999 |
TISAB II for Fluoride Electrode | 4 x 1 Gal. |
£436.27
Pack of 4 |
In Stock
Log in to see stock. |
||||
11777777
|
Thermo Scientific™
941609 |
Sulfide Anti-Oxidant Buffer (SAOB) for Sulfide Electrode | 4 x 475 mL |
£275.36
475mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11747757
|
Thermo Scientific™
941709 |
Complextion Ionic Strength Adjuster (CISA) for Chloride Electrode | 2 x 1 L |
£292.57
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10598741
|
Thermo Scientific™
950210 |
ISA for Carbon Dioxide Electrode | 475 mL |
£104.12
475mL |
In Stock
Log in to see stock. |
||||
11777677
|
Thermo Scientific™
951011 |
Alkaline Reagent for Cyanide Electrode | 475 mL |
£104.98
475mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12993247
|
Thermo Scientific™
951210 |
pH Adjusting Low Level ISA for Ammonia Electrode | 475 mL |
£104.98
475mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12881521
|
Thermo Scientific™
951211 |
pH Adjusting ISA for Ammonia Electrode | 475 mL |
£97.24
475mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12903257
|
Thermo Scientific™
951213 |
Storage Solution for Ammonia Electrode | 475 mL |
£63.25
475mL |
In Stock
Log in to see stock. |
||||
11767687
|
Thermo Scientific™
956410 |
Acid Buffer for Nitrogen Oxide Electrode | 475 mL |
£111.86
475mL |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
11767807
|
Thermo Scientific™
977010 |
Iodide Reagent for Residual Chlorine Electrode | 5 x 50 mL |
£97.24
Pack of 5 |
In Stock
Log in to see stock. |
||||
11737707
|
Thermo Scientific™
977011 |
Acid Reagent for Residual Chlorine Electrode | 475 mL |
£84.16
475mL |
In Stock
Log in to see stock. |
||||
Description
Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.
Warranty: Out of box
Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements. The ammonia pH-adjusting ISA (951211) and low-level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low-level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.
All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:
- Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
- Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
- Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
- Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
Specifications
Ion Selective Electrode | |
Ion Selective Electrode Reagents | |
Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.