missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions

Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).

Brand:  Thermo Scientific™ 654203

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11792737

  • £78.39 / Each
Estimated Shipment: 24-09-2024
to see stock.

Description

Description

Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).

All surfactant analysis solutions are packaged as one 1 pint (475mL) bottle, except Cat. No. 654202 packaged as one 2 oz. (60mL) bottle.
Specifications

Specifications

Triton X-100 Sample Additive
Triton X-100 Sample Additive, 475mL
475 mL
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.