Learn More
Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions
Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).
Brand:  Thermo Scientific™ 654201
Additional Details : CAS Number : 121-54-0 Weight : 0.47700kg
 View more versions of this product
Product Code. 12879201
  • £69.79 / 475mL
In Stock
to see stock.
Description
Description
Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).
All surfactant analysis solutions are packaged as one 1 pint (475mL) bottle, except Cat. No. 654202 packaged as one 2 oz. (60mL) bottle.
Specifications
Specifications
Hyamine 1622 Titrant, 475mL
475 mL
Bottle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.