Learn More
Thermo Scientific™ Orion Lab Star PH111 Bench pH Meters
Description
Discover the award-winning Orion Lab Star PH111 pH/mV bench meter. Measure pH or ORP simultaneously with mV and temperature. Featuring a large, backlit display for easy viewability and small footprint to save bench space, this pH meter is ideal for simplified, budget-friendly lab testing. Use the onscreen stabilizing and ready indicators to help identify when pH meter and ORP meter readings are complete. View active pH, ORP and temperature calibration logs with date and time stamp, record key measurements using the 500-point data log with date and time stamp, and output all logs to a printer or computer.
Orion Lab Star PH111 pH Meter Features
- Budget-friendly pH meter for simplified pH and ORP testing
- Display shows pH, mV and temperature simultaneously for accurate data recording
- Measure -2.000 to 18.000 pH, -2000.0 to +2000.0 mV and -5.0 to 105.0 °C or 23.0 to 221.0 °F to ensure comprehensive measurement capabilities
- Perform one to five point pH calibrations, one point ORP calibrations and one point temperature adjustment calibrations for proficient SOP compliance
- Large 5 inch backlit display makes viewing and recording measurements easy
- Small meter footprint saves valuable bench space in your lab
- Onscreen stabilizing and ready indicators help identify when readings are complete
- pH electrode condition icon helps ensure reliable electrode performance
- Date and time data stamped data and calibration logs support GLP compliance
- View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements
- Use 500-point data log with date and time stamp to record key measurements
- Output data and calibration logs to a printer or computer for long-term data storage
Meter and Meter Kits
- LSTAR1110 is the pH meter and stand option, which allows you to purchase your pH electrode separately
- LSTAR1115 is the standard pH meter kit option, for common pH samples such as drinking water, tap water, buffers (non-TRIS) and general purpose student use
- LSTAR116 is the easy-to-clean bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included refillable pH electrode is easy-to-clean and restore
- LSTAR117 is the low-maintenance bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included gel-filled pH electrode is low-maintenance and easy to operate
- LSTAR1118 is the difficult sample pH meter kit option, for pH samples that are thick, viscous or semi-solid including foods, beverages, wastewaters, emulsions, soils, slurries, suspended solids or sludges
Specifications
Specifications
| Description | Orion Lab Star PH111 bench pH/mV meter difficult sample kit |
| Includes | 9165 BNWP Sure-Flow refillable epoxy-body pH electrode 927007MD stainless steel ATC temperature probe 916099 pH 4/7/10 buffer, storage and cleaning solution 60 mL kit meter with electrode stand, PC cable, universal power adapter |
| Accuracy (Temperature) | ±0.3°C/±0.5°F |
| Accuracy (mV) | ±0.2 or ±0.05%, whichever is greater |
| Accuracy (pH) | ±0.005, ±1 LSD |
| Data Management Details | Transfer single or all data points to printer or computer |
| Outputs | Computer or printer (STARA-106) |
| For Use With (Application) | Difficult Sample pH Meter Kit Option, for pH Samples That Are Thick, Viscous or Semi-Solid Including Foods, Beverages, Wastewaters, Emulsions, Soils, Slurries, Suspended Solids or Sludges |
| Measurement Modes | pH with mV and Temperature, ORP with mV and Temperature |
| Calibration | pH, ORP and temperature |
| Vedi altri risultati |
Sicurezza e movimentazione
- Lab Star PH111 pH/mV bench meter difficult sample
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
- P321-Specific treatment (see supplemental first aid instructions on this label).
- MIXTURE LIST-Contains : potassium chloride,silver nitrate, triton X-100, sodium lauryl sulphate
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.