missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ORP Standards for Redox/ORP Electrodes

Product Code. p-4521270
Click to view available options
Quantity:
475 mL
5 x 60 mL
Unit Size:
475mL
Pack of 5
This item is not returnable. View return policy

Product Code. 11757807

Brand: Thermo Scientific™ 967961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measure ORP simply and accurately with these standards, stable for up to a year (unopened).

Measure ORP simply and accurately with Thermo Scientific™ Orion™ ORP Standards for Redox/ORP Electrodes. Orion OPR standards are stable for up to a year (unopened) and can be used directly from the bottle with no mixing or diluting.

Orion OPR standards are designed for use with gel-filled ORP/ATC Triode™ electrodes (9179BNMD and 9179BN); refillable ORP/ATC Triode electrodes (9180BNMD and 9180BN); glass-bodied ORP electrodes, (9778BNWP and 9778SC); and Sure-Flow™ ORP electrodes, (9678BNWP and 967800).

  • Included in the new ASTM ORP method
  • Used as relative mV standard to calibrate the offset of sample
  • +220mV vs. Ag/AgCl using 4M KCl saturated with AgCl filling solution (Part No. 900011), +420mV vs. Standard Hydrogen Electrode (EH)
TRUSTED_SUSTAINABILITY

Specifications

Description ORP Standard
Packaging Bottle
For Use With (Equipment) Gel-filled ORP/ATC Triode™ electrodes (9179BNMD and 9179BN), Refillable ORP/ATC Triode electrodes (9180BNMD and 9180BN), Glass-bodied ORP electrodes, (9778BNWP and 9778SC), Sure-Flow™ ORP electrodes, (9678BNWP and 967800)
Type Redox/ORP Electrode Standard
Quantity 5 x 60 mL
compliance-icons compliance-icons
Product Identifier
  • ORP Standard
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360-May damage fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P321-Specific treatment (see supplemental first aid instructions on this label).
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,boric acid

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.