Learn More
Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.
Brand: Thermo Scientific™ 910725
41.69 GBP valid until 2024-12-31
Use promo code "21980" to get your promotional price.
Description
Ensure fresh, ready-to-use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches. Buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.
- Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
- Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- Pouches are 15mL each and are available in 10 packs or 25 packs
Note:
The following part numbers have been changed due to a recent system upgrade.
Old Part Number: 910410
New Part Number: 910410-WA
Old Part Number: 911025
New Part Number: 911025-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Specifications
pH Buffer, individual use pouches | |
Light Yellow | |
pH calibration | |
7 | |
25 x 15 mL | |
Water, Dibasic, Potassium Dihydrogen Phosphate, 5-Bromo-5-Nitro-1,3-Dioxane, FD & C Yellow #5, Sodium Phosphate | |
Yes |
100°C | |
pH 7.00 Buffer | |
pH Meter and Electrode, single-use | |
Liquid | |
Buffer | |
Pouch | |
Soluble |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.