missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches

Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.

Brand:  Thermo Scientific™ 911025-WA

42.36 GBP valid until 2024-12-31
Use promo code "21980" to get your promotional price.


 View more versions of this product

Product Code. 10470443

  • £44.67 / Pack of 25

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Ensure fresh, ready-to-use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches. Buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.

  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs

Note:

The following part numbers have been changed due to a recent system upgrade.

Old Part Number: 910410

New Part Number: 910410-WA

Old Part Number: 911025

New Part Number: 911025-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

pH Buffer, individual use pouches
Light Blue
pH calibration
10.01
25 x 15 mL
Water, Sodium Bicarbonate, Methylparaben, FD & C Blue #1, Sodium Carbonate
Yes
100°C
pH 10.01 Buffer
pH Meter and Electrode, single-use
Liquid
Buffer
Pouch
Soluble
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.