missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches

Produktkod. 10756861
Klicka för att se tillgängliga alternativ
Quantity:
10 x 15 mL
25 x 15 mL
pH:
4.01
7
10.01
Förpackningsstorlek:
Pack of 10
Pack of 25
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 10756861

Brand: Thermo Scientific™ 910425

Please to purchase this item. Need a web account? Register with us today!

Edge
Denna artikel kan inte returneras. Se returpolicy

Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.

Ensure fresh, ready-to-use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches. Buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.

  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs

Note:

The following part numbers have been changed due to a recent system upgrade.

Old Part Number: 910410

New Part Number: 910410-WA

Old Part Number: 911025

New Part Number: 911025-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material pH Buffer, individual use pouches
Boiling Point 100°C
Color Light Red
Description pH 4.01 Buffer
For Use With (Application) pH calibration
For Use With (Equipment) pH Meter and Electrode, single-use
pH 4.01
Physical Form Liquid
Quantity 25 x 15 mL
Solution Type Buffer
Formulation Water, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate
Packaging Pouch
Traceability to NIST Yes
Solubility Information Soluble
Visa mer Visa mindre

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.