Learn More
Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.
£26.88 - £44.67
Specifications
Chemical Name or Material | pH Buffer, individual use pouches |
---|---|
Boiling Point | 100°C |
For Use With (Application) | pH calibration |
For Use With (Equipment) | pH Meter and Electrode, single-use |
Physical Form | Liquid |
Product Code | Brand | Quantity | pH | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | pH | Price | Quantity & Availability | ||||
10283061
|
Thermo Scientific™
910410-WA |
10 x 15 mL | 4.01 |
£26.88
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10756861
|
Thermo Scientific™
910425 |
25 x 15 mL | 4.01 |
£44.67
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10069110
|
Thermo Scientific™
910710 |
10 x 15 mL | 7 |
£26.88
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10242491
|
Thermo Scientific™
910725 |
25 x 15 mL | 7 |
£44.67
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10185062
|
Thermo Scientific™
911010 |
10 x 15 mL | 10.01 |
£26.88
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10470443
|
Thermo Scientific™
911025-WA |
25 x 15 mL | 10.01 |
£44.67
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ensure fresh, ready-to-use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches. Buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.
- Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
- Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- Pouches are 15mL each and are available in 10 packs or 25 packs
Note:
The following part numbers have been changed due to a recent system upgrade.
Old Part Number: 910410
New Part Number: 910410-WA
Old Part Number: 911025
New Part Number: 911025-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Warranty: Out of box
Specifications
pH Buffer, individual use pouches | |
pH calibration | |
Liquid | |
Pouch | |
Soluble |
100°C | |
pH Meter and Electrode, single-use | |
Buffer | |
Yes |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.