Learn More
Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions
Product Code. 11772168
Click to view available options
Filling Solution Type:
A, B, C, D
Bacteria Removal (B)
General Cleaning (C)
No Cal Stain Lifter
Oil and Grease Removal (D)
Protein Removal (A)
Rinse
Quantity:
10 pk.
25 pk.
4 x 30 mL
4 x 60 mL
60 mL
60 mL (B, D), 30 mL (A, C)
Unit Size:
60mL
Pack of 10
Pack of 25
Pack of 4
Product Code. 11772168
Brand: Thermo Scientific™ 911110
Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.
Cleaning Solutions:
  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
TRUSTED_SUSTAINABILITY
Specifications
Description 10 Pack of pH Electrode Rinse Solution Pouches
For Use With (Equipment) pH Electrode
Type Rinse Solution
Filling Solution Type Rinse
For Use With (Application) Solutions restore proper electrode performance and prolong the useful life of the electrode
Packaging Pouch
Quantity 10 pk.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.