Learn More
Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.
Brand: Thermo Scientific™ 900021-WA
Code : NEW
Additional Details : Weight : 0.19710kg
Description
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.
Cleaning Solutions:
- Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
- Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
Specifications
4 x 1 oz. (4 x 30mL) Bottles of Cleaning Solution A for Protein Removal, Beaker and Pipet | |
pH Electrode Cleaning Solution | |
30 mL |
pH Electrode | |
Cleaning |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.