missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions

Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.

£29.94 - £66.95

Specifications

For Use With (Equipment) pH Electrode
View More Specs

Products 9
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11722737
View Documents Promotion Details
Thermo Scientific™
510015
2 oz. (60mL) Bottle of No Cal Stain Lifter Solution
£29.94
60mL
View alternative products
Save Up to 
»
Estimated Shipment: 23-07-2024
to see stock.
Add to basket
 
11772787
View Documents Promotion Details
Thermo Scientific™
900020
1 Bottle Each of Cleaning Solutions A, B, C, and D; Beaker and Pipet
£57.22
Pack of 4
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11727777
View Documents Promotion Details
Thermo Scientific™
900021-WA
4 x 1 oz. (4 x 30mL) Bottles of Cleaning Solution A for Protein Removal, Beaker and Pipet
£62.39
Pack of 4
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11737777
View Documents Promotion Details
Thermo Scientific™
900022-WA
4 x 2 oz. (4 x 60mL) Bottles of Cleaning Solution B for Bacteria Removal, Beaker and Pipet
£66.95
Pack of 4
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11747777
View Documents Promotion Details
Thermo Scientific™
900023
4 x 1 oz. (4 x 30mL) Bottles of Cleaning Solution C for General Cleaning, Beaker and Pipet
£63.68
Pack of 4
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11757777
View Documents Promotion Details
Thermo Scientific™
900024
Solution D 4 x 2 oz. (4 x 60mL) Bottles of Cleaning Solution D for Oil and Grease Removal, Beaker and Pipet
£63.68
Pack of 4
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
11772168
View Documents Promotion Details
Thermo Scientific™
911110
10 Pack of pH Electrode Rinse Solution Pouches
£37.95
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
11702188
View Documents Promotion Details
Thermo Scientific™
911125
25 Pack of pH Electrode Rinse Solution Pouches
£52.83
Pack of 25
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
 
15304338
View Documents Promotion Details
Thermo Scientific™
900021-WA
4 x 1 oz. (4 x 30mL) Bottles of Cleaning Solution A for Protein Removal, Beaker and Pipet This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.

Cleaning Solutions:

 

  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for removing bacterial contaminants, cleaning solution C is for general cleaning and cleaning solution D is for removing oil and grease contaminants
  • Bottles include a small beaker to hold the solution and a plastic pipet to remove the old electrode filling solution

Includes (PN 900020):

  • One bottle each of cleaning solutions A, B, C and D
  • One beaker
  • One pipet

Note:

The following part numbers have been changed due to a recent system upgrade:

Old Part Number: 900021

New Part Number: 900021-WA

Old Part Number: 900022

New Part Number: 900022-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.

Warranty: Out of box


Specifications

Specifications

pH Electrode
Videos
SDS
Documents

Documents

Product Certifications

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.