Learn More
Thermo Scientific™ Orion™ Potassium Electrodes
Measure potassium ions in aqueous solutions simply, accurately and economically.
Brand:  Thermo Scientific™
630.48 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 10410643
  • £749.27 / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Measure potassium ions in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Potassium Electrodes.
  • Calibration Standard Catalog number: 0.1M KCl / 921906
  • Required ISA Catalog number: 931911
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BNC
drift-free potentials
ISE Meters
Potassium Half Cell Electrode with BNC connector
Free-flowing Sure-Flow junction assures stable
Combination Ion Selective Electrode