Learn More
Thermo Scientific™ Orion™ Powder Chemistry Packs
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Brand: Thermo Scientific™ AC4P82
40.22 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
View more versions of this product
Description
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
- Equivalent to other widely used and recognized chemistries
- Compatible with Orion™ AQUAfast™ Colorimeters
Specifications
AQUAfast sulfate reagent powder packs, 100 tests | |
Drinking Water: ASTM D516-07 / Standard Methods 4500-SO42- E; Wastewater: ASTM D516-07 / Standard Methods 4500-SO42- E | |
100 Sulfate Powder Packs | |
100 tests | |
AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Sulfate | |
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter | |
Powder Pack | |
5 to 100 mg/L SO4 | |
Bariumsulfate-Turbidity |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : citric acid, bariumchloride dihydrate
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.