Learn More
Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes
Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance with these redox/ORP/temp electrodes.
£298.59 - £417.06
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
10691033
|
Thermo Scientific™
9179BN |
Epoxy Triode Low Maintenance | Orion A+ Series Meters |
£298.59
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11732857
|
Thermo Scientific™
9179BNMD |
Epoxy Triode Low Maintenance | Orion Star Series Meters |
£325.26
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
10080514
|
Thermo Scientific™
9180BNMD |
Epoxy Refillable Triode enance | Orion Star Series Meters |
£400.13
Each |
In Stock
Log in to see stock. |
||||
10315173
|
Thermo Scientific™
9180BN |
Epoxy Refillable Triode | Orion A+ Series Meters |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance. Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes measure ORP and temperature using a platinum redox sensor, a silver/silver chloride internal reference system and a built-in thermistor which is adapted for Automatic Temperature Compensation (ATC).
Features:- Available in epoxy triode low maintenance and epoxy refillable triode construction
- Built-in thermistor for Automatic Temperature Compensation (ATC)
- Not designed for amperometric or constant current titrations, such as Karl Fischer analysis
Specifications
Epoxy | |
Redox/ORP/Temp Electrode |