Learn More
Thermo Scientific™ Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Perform virtually any electrochemistry analysis with these conductivity standards, solutions and accessories.
£552.63 - £635.69
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11708216
|
Thermo Scientific™
013017 |
Replacement Glass Flow Cell | N/A | ||||||
11732637
|
Thermo Scientific™
1010001 |
Conductivity Calibration Resistor Kit with 8 Pin MiniDIN Connection |
£552.63
Each |
In Stock
Log in to see stock. |
|||||
Description
Confidently perform conductivity tests and measurements with Thermo Scientific™ Orion™ Accessories. Find an additional or replacement Glass Flow Cell for use with the Orion 2-Cell Ultra-pure Conductivity Probe. Use the Thermo Scientific Orion Conductivity Resistor Calibration Kit to verify the performance of Thermo Scientific™ Orion™ Versa Star Pro™ and Orion Star™ Bench and Portable Conductivity Meters. Each conductivity calibration kit includes six resistors that simulate nominal conductance values between 1 μS and 100 mS and have 8 pin MiniDIN connectors.
- Traceable to NIST standard reference materials
- Designed to meet all end user requirements, including laboratory grade and DIN standards
- Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
- Glass Flow Cell compatible with Thermo Scientific™ Orion™ Orion 2-cell Ultrapure Conductivity Probe, (Cat. No 013016MD)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.