Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™
Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
£372.09 - £375.52
Specifications
Junction Ceramic
Material Chemical Resistant Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type pH/ATC Combination Electrode
View More Specs
Products 2
Product Code Brand Connector Types For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Connector Types For Use With (Equipment) Price Quantity & Availability  
16690713
View accessory products View Documents Promotion Details
Thermo Scientific™
8302BNURCA
BNC Waterproof, RCA Mettler Toledo Five and Seven series pH meters
£372.09
Each
In Stock
to see stock.
Add to basket
12493617
View accessory products View Documents Promotion Details
Thermo Scientific™
8302BNUMD
BNC Waterproof, 8-pin MiniDIN Orion Versa Star, Star A and Dual Star Meters
£375.52
Each
In Stock
to see stock.
Edge
Add to basket
Description
Show More
Specifications
Specifications
Ceramic
0 to 14
pH/ATC Combination Electrode
ROSS
0.03 pH for buffers using automatic temperature compensation
2 Years
1 m
Chemical Resistant Glass Body
0°C to 100°C
ROSS electrode 3 M KCl fill solution (Cat. No. 810007)
7
0.01
120 mm
12 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™