missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Epoxy Bodied pH/ATC Triode™

Product Code. 11839221
Click to view available options
Connector Types:
BNC (Waterproof), 8-pin Mini-DIN
BNC (Waterproof), RCA
Length (Metric) Cable:
1 m
3 m
Unit Size:
Each

Product Code. 11839221

Brand: Thermo Scientific™ 8107BNUMD

Only null left
Add to basket
Add to basket

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.

Thermo Scientific™ Orion™ ROSS Ultra Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra line with a built-in ATC. These products offer a low-maintenance, gel-filled, epoxy body and are best used where minimal maintenance is required. Non-refillable Orion ROSS Ultra Triodes come with an 18 month warranty.
 

Measure pH and Temperature with One Low-Maintenance Electrode

Low ionic strength samples, such as clean waters, present a challenge to pH electrode performance. Sluggish response, poor accuracy and drift are common. The Thermo Scientific Orion ROSS Ultra 8107 Triode pH electrode responds within one minute, proving a stable, accurate reading.

Complete your pH measurements faster. Get long, maintenance-free electrode lifetime with special long-life gel and innovative capillary glass junction design. Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.

Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.

Target Markets:

  • Pharmaceutical/Biopharmaceutical
  • Food Manufacturing
  • Academic/Government
  • Environmental
Applications:
  • Quality Control
  • Research
  • General Purpose
TRUSTED_SUSTAINABILITY

Specifications

Description Orion ROSS Ultra Low Maintenance pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 1 meter cable
Connector Types BNC (Waterproof), 8-pin Mini-DIN
Junction Glass Capillary
Material Rugged Epoxy Body, Filled with Low Maintenance Gel
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type pH/ATC Combination Electrode
Filling Solution Type None required
For Use With (Equipment) Orion Versa Star, Star A and Dual Star Meters
Internal Reference ROSS
Isopotential Point 7
pH Relative Accuracy 0.03 pH for buffers using automatic temperature compensation
Precision 0.01
Sample Compatibility TRIS, sulfides, or proteins
Warranty 18 Months
Length (Metric) 120 mm
Length (Metric) Cable 1 m
Diameter (Metric) 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.