missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes

Measure both pH and temperature in samples with these low maintenance, pH/ATC combination electrodes.

Brand:  Thermo Scientific™ 8107UWMMD

 View more versions of this product

Product Code. 11849221

  • £354.52 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Increase the accuracy of your readings with the Thermo Scientific™ Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes. These combination electrodes have a pH sensor that delivers superior performance at varying sample temperatures. They combine the stability, precision and speed of response of the ROSS Ultra line with integrated temperature measurement. Orion ROSS electrodes contain no silver or silver chloride and incorporate a double-junction design, which protects the sample from contamination and makes the electrodes compatible with TRIS buffers.

Measure pH and Temperature with One Low-Maintenance Electrode

Low ionic strength samples, such as clean waters, present a challenge to pH electrode performance. Sluggish response, poor accuracy and drift are common. The Thermo Scientific Orion ROSS Ultra 8107 Triode pH electrode responds within one minute, proving a stable, accurate reading.

Complete your pH measurements faster. Get long, maintenance-free electrode lifetime with special long-life gel and innovative capillary glass junction design. Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.

Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.

Target Markets:

  • Pharmaceutical/Biopharmaceutical
  • Food Manufacturing
  • Academic/Government
  • Environmental
Applications:
  • Quality Control
  • Research
  • General Purpose
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orion ROSS Ultra Low Maintenance pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 3 meter cable
Glass Capillary
0 to 14
pH/ATC Combination Electrode
Orion Versa Star, Star A and Dual Star Meters
7
0.01
18 Months
3 m
BNC Waterproof, 8-pin MiniDIN
Rugged Epoxy Body, Filled with Low Maintenance Gel
0°C to 80°C
None required
ROSS
0.03 pH for buffers using automatic temperature compensation
TRIS, sulfides, or proteins
120 mm
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.