Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes
Measure both pH and temperature in samples with these low maintenance, pH/ATC combination electrodes.
£342.47 - £366.00
Specifications
Junction | Glass Capillary |
---|---|
Material | Rugged Epoxy Body, Filled with Low Maintenance Gel |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 80°C |
Type | pH/ATC Combination Electrode |
Product Code | Brand | Connector Types | Length (Metric) Cable | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connector Types | Length (Metric) Cable | For Use With (Equipment) | Price | Quantity & Availability | |||
11839221
|
Thermo Scientific™
8107BNUMD |
BNC Waterproof, 8-pin MiniDIN | 1 m | Orion Versa Star, Star A and Dual Star Meters |
£342.47
Each |
In Stock
Log in to see stock. |
|||
11849221
|
Thermo Scientific™
8107UWMMD |
BNC Waterproof, 8-pin MiniDIN | 3 m | Orion Versa Star, Star A and Dual Star Meters |
£354.52
Each |
In Stock
Log in to see stock. |
|||
15950377
|
Thermo Scientific™
8107BNURCA |
BNC Waterproof, RCA | 1 m | Mettler Toledo Five and Seven Series pH Meters |
£366.00
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
Description
Increase the accuracy of your readings with the Thermo Scientific™ Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes. These combination electrodes have a pH sensor that delivers superior performance at varying sample temperatures. They combine the stability, precision and speed of response of the ROSS Ultra line with integrated temperature measurement. Orion ROSS electrodes contain no silver or silver chloride and incorporate a double-junction design, which protects the sample from contamination and makes the electrodes compatible with TRIS buffers.
Measure pH and Temperature with One Low-Maintenance Electrode
Low ionic strength samples, such as clean waters, present a challenge to pH electrode performance. Sluggish response, poor accuracy and drift are common. The Thermo Scientific Orion ROSS Ultra 8107 Triode pH electrode responds within one minute, proving a stable, accurate reading.
Complete your pH measurements faster. Get long, maintenance-free electrode lifetime with special long-life gel and innovative capillary glass junction design. Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.
Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.
Target Markets:
- Pharmaceutical/Biopharmaceutical
- Food Manufacturing
- Academic/Government
- Environmental
Applications:
- Quality Control
- Research
- General Purpose
Warranty: 18 months from date of purchase by Buyer or 24 months from date of shipment from Seller, whichever is earlier.
Specifications
Glass Capillary | |
0 to 14 | |
pH/ATC Combination Electrode | |
ROSS | |
0.03 pH for buffers using automatic temperature compensation | |
18 Months | |
12 mm |
Rugged Epoxy Body, Filled with Low Maintenance Gel | |
0°C to 80°C | |
None required | |
7 | |
0.01 | |
120 mm |