Learn More
Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter
Connect a USB port to the RS232 port of your meter using the Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter.
Brand: Thermo Scientific™ 1010005
Code : 59
Additional Details : Weight : 1.00000kg
Description
Connect a USB port to the RS232 port of your meter using the Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter. This adapter allows interface between a USB connection and Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion™ Star A series (Star A320, Star A220 and Star A210 series) meters and legacy Orion™ Star and Star Plus meters.
Specifications
Orion Star series RS232 to USB adapter cable | |
Meter Interface Cable | |
RS232 to USB computer interface adapter and cable (2.5mm RS232 meter connector and USB computer connector) |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.