Learn More
Thermo Scientific™ Orion™ Sodium Electrodes
Measure sodium in aqueous solution quickly, accurately and economically with these sodium electrodes.
Brand: Thermo Scientific™ 8411BN
389.56 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : 59
Additional Details : Weight : 3.06900kg
Description
Measure sodium in aqueous solutions quickly, accurately and economically with Thermo Scientific™ Orion™ Sodium Electrodes. A unique redox internal system with a Thermo Scientific™ Orion™ ROSS™ electrode provides faster response, and greater stability and accuracy than conventional sodium electrodes.
8611BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. 8411BN is a half cell sensor that requires ROSS reference electrode, Cat. No. 800300 or the ROSS Ultra reference electrode, Cat. No. 800500U to function.
The sodium electrode is commonly used to measure samples such as food, beverages and animal feed.
Specifications
Measures sodium in aqueous solution quickly, accurately, and economically; Unique redox internal system with ROSS™ electrode provides faster response, greater stability and accuracy than conventional sodium electrodes | |
ROSS half-cell BNC Orion sodium electrode | |
13 mm | |
800300 or 800500U | |
1M | |
40 in | |
Saturated to 10-6 M, saturated to 0.02ppm | |
ISE Meters | |
1 x 10-6 M to saturated 0.02 ppm to saturated | |
No |
BNC | |
0.47 in | |
900010 for regular Na+ (> 10-5 M or 0.2 ppm) 900012 for low level Na+ (< 10-5 M or 0.2 ppm) |
|
4.72 in | |
121 mm | |
Glass Body | |
32 to 176°F | |
0°C to 100°C | |
Sodium Half-Cell Ion Selective Electrode |
Safety and Handling
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362-Take off contaminated clothing.
- P403-Store in a well-ventilated place.
- P501-Dispose of contents/container to .