Learn More
Thermo Scientific™ Orion™ Star A Series Benchtop Meter-Attached Electrode Stand
Use the Thermo Scientific™ Orion™ Star A Series Benchtop Meter-Attached Electrode Stand to easily place sensors in samples, simplify rinsing and storage and minimize breakage
Brand: Thermo Scientific™ STARA-BEA
Code : 59
Additional Details : Weight : 0.27900kg
Description
Use the Thermo Scientific™ Orion™ Star A Series Benchtop Meter-Attached Electrode Stand to easily place sensors in samples, simplify rinsing and storage between samples and minimize electrode breakage using the stand's cable management system. Attach the stand to either side of Thermo Scientific™ Orion™ Versa Star benchtop meters and Thermo Scientific™ Orion™ Star A210 and Star A110 series benchtop meters or use the stand with weighted base (Cat. No. STARA-HB) for compatibility with any meter system. Includes meter bracket, arm and electrode holder (Cat. No. STARA-ESCD).
Specifications
Orion Star A series meter stand, electrode holder and meter-attachable base | |
Electrode Stand/Holder | |
Redesigned electrode holder (Cat .No. STARA-ELHD), plastic stand with cable management system and meter-attachable base | |
To use with almost any Orion benchtop or portable meter, purchase the weighted base separately (Cat. No. STARA-HB) |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.