Learn More
Thermo Scientific™ Orion Star™ A211 Benchtop pH Meter

Description
Record accurate and reliable pH, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A211 Benchtop pH Meter. This meter is ideal for a wide range of applications and advanced pH analysis in the lab. Perform up to a 5 point pH calibration, log up to 2000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
Thermo Scientific Orion Star A211 Benchtop pH Meters are easy-to-use, dependable laboratory meters with advanced features and functions. Selectable read modes and stability indicator give you flexibility and control over your testing. All critical data is shown with your measurements including temperature, date and time, read type, measurement stability, active calibration information and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates.
Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view and stirrer keys that ensure easy operation. All setup menus are in one location with direct access using a single shortcut key so you can quickly access, evaluate and change measurement and instrument settings.
- Measure pH, mV, relative mV or ORP with temperature
- Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
- Calibration editing option allows individual points to be fixed without a full recalibration
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe (catalog number 096019) to uniformly mix solutions
- Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
- Calibration log stores 10 most recent calibrations
- Non-volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with universal power adapter or optional AA batteries
- IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
- Meters include CE, TUV 3-in-1 and FCC Class A certifications
- 3-year replacement warranty
Specifications
Specifications
| Description | Orion Star A211 pH benchtop meter durable sensor TRIS kit with TRIS-compatible ROSS Ultra epoxy-body pH/ATC electrode |
| Includes | Star A211 pH meter, 8157BNUMD ROSS Ultra epoxy-body pH/ATC electrode, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable |
| Accuracy (Temperature) | ±0.1°C |
| Accuracy (mV) | ±0.2 or ±0.05%, whichever is greater |
| Accuracy (pH) | ±0.002 |
| Data Management Details | Transfer single, range or all data points to printer or computer |
| Outputs | USB, RS-232 |
| Measurement Modes | pH, mV, Relative mV, ORP with Temperature |
| Calibration | pH with calibration editing option, relative mV, ORP and temperature |
| Range (pH) | -2 to 20 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.