missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion Star T900 Series Titrator Accessories

Minimize maintenance downtime with easy to install Orion Star T900 series titrator replacement parts. Orion Star T900 series titrator accessories expand functionality.

Brand:  Thermo Scientific™ START-B50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15802117

  • £360.54 / Each
In Stock
to see stock.
Add to basket

Description

Description

Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.

•Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
•Bottle holder helps secure titrant to prevent spills and offers flexible placement
•Color-coded tubing and connectors simplify tubing installation and maintenance
•Computer software is designed to make data transfer simple and reliable

Specifications

Specifications

Orion Star T900 series 50ml burette
Titrator accessory, burette
Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Product Certifications
Ver productos accesorios
Ofertas especiales

Ofertas especiales

Garantía