Learn More
Thermo Scientific™ Orion Star T900 Series Titrator Accessories
Minimize maintenance downtime with easy to install Orion Star T900 series titrator replacement parts. Orion Star T900 series titrator accessories expand functionality.
Brand: Thermo Scientific™
Description
Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.
•Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
•Bottle holder helps secure titrant to prevent spills and offers flexible placement
•Color-coded tubing and connectors simplify tubing installation and maintenance
•Computer software is designed to make data transfer simple and reliable
Specifications
Orion Star T900 series 1L plastic bottles, pack of 12 | |
Titrator accessory, reagent bottle | |
Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.