missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Storage Sleeve and Base

Protect electrodes from breakage when not in use with the Thermo Scientific™ Orion™ Electrode Storage Sleeve and Base.

Brand:  Thermo Scientific™ 810017

37.61 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 13400789

  • £40.27 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Protect electrodes from breakage when not in use with the Thermo Scientific™ Orion™ Electrode Storage Sleeve and Base.

The Thermo Scientific Orion Storage Sleeve and Base is for use with pH electrodes that are 120mm or less in length that have a diameter of 12mm. The sleeve protects your electrode from breakage when not in use and will not tip, due to the weighted removable base. As an additional convenience, the pH bulb stays conditioned while in storage so that it is ready to use when removed from the sleeve.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orion Storage Sleeve and Base
pH Electrode
Storage Sleeve, Base
Product Suggestions

Product Suggestions

Videos
SDS
Documents Product Certifications
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Orion™ Storage Sleeve and Base > Storage sleeve and base

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.