missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Total Alkalinity Test Kit

Product Code. p-4521254
Click to view available options
Quantity:
475 mL
4 x 475 mL
Type:
Alkalinitiy Reagent Refill
Alkalinity Standard/Control Refill
Total Alkalinity Test Kit
Unit Size:
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 10140971

Brand: Thermo Scientific™ 700010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Save time by eliminating additional equipment set-up, titrations and calculations with this total alkalinity test kit that provides two-step direct measurement.

Save time by eliminating additional equipment set-up, titrations and calculations. The Thermo Scientific™ Orion™ Total Alkalinity Test Kit provides two-step direct measurement of total alkalinity and pH using a pH electrode.

Features:

  • Direct reading of total alkalinity from 0 to 200ppm as CaCO3, from the enclosed pH conversion chart
  • No interference from color or turbidity

Includes:

  • One 1 pint (475mL) bottle alkalinity reagent (700011)
  • One 1 pint (475mL) bottle alkalinity standard/control (700012)
  • Total alkalinity conversion chart
  • Application procedure No. 517, Total Alkalinity in Natural Waters

Note:

The following part number has been changed due to a recent system upgrade.

Old Part Number: 700011

New Part Number: 700011-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Data Safety Sheet (MSDS) files.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Composition Non Hazardous Media
Format Liquid
For Use With (Application) two-step direct measurement of total alkalinity and pH using a pH electrode; Saves time by eliminating additional equipment set-up, tedious titrations and calculations
For Use With (Equipment) pH Electrode
Includes 475mL Alkalinity Reagent (700011), 475mL Alkalinity Standard/Control (700012), Total Alkalinity Conversion Chart and Application Procedure No. 517, Total Alkalinity in Natural Waters
Packaging Kit
Quantity 475 mL
Type Total Alkalinity Test Kit

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.