Learn More
Thermo Scientific™ Orion™ Versa Star Pro™ pH/ISE Benchtop Multiparameter Meter
Get premium performance and ultimate flexibility with our benchtop meter with pH/ISE module for pH, ion concentration, and temperature.
£1660.74 - £2460.99
Specifications
Outputs | USB (2), RS232 |
---|---|
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR) |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
11671499
|
Thermo Scientific™
VSTAR40A |
Orion Versa Star Pro benchtop meter with pH/ISE/mV module and stand | Versa Star Pro meter, VSTAR-ISE pH/ISE module, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
£1660.74
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
11681499
|
Thermo Scientific™
VSTAR40A2 |
Orion Versa Star Pro benchtop meter kit with pH/ISE/mV module, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe, solutions and stand | Versa Star Pro meter, VSTAR-ISE pH/ISE module, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
£1961.91
Each |
In Stock
Log in to see stock. |
||||
11691499
|
Thermo Scientific™
VSTAR40B |
Orion Versa Star Pro benchtop meter with two pH/ISE/mV modules and stand | Versa Star Pro meter, two VSTAR-ISE pH/ISE modules, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
£2142.61
Each |
In Stock
Log in to see stock. |
||||
11601509
|
Thermo Scientific™
VSTAR40B2 |
Orion Versa Star Pro benchtop meter kit with two pH/ISE/mV modules, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe, solutions and two stands | Versa Star Pro meter, two VSTAR-ISE pH/ISE modules, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
£2460.99
Each |
In Stock
Log in to see stock. |
||||
Description
Get high accuracy, premium performance and ultimate flexibility with the Thermo Scientific™ Orion™ Versa Star Pro™ pH/ISE Benchtop Multiparameter Meter. Meet your most challenging applications for pH, ion concentration, mV, ORP and temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. Perform up to 6 point pH and ISE calibrations, log up to 2000 data point sets with time/date stamp and transfer calibration and data logs via USB or RS232 to a printer or computer. Select systems with one or two pH/ISE modules.
- Onscreen measurement stability indicator lets you know exactly when your measurements are ready
- Selectable read modes include Auto-Read to hold a stable reading, Continuous to show changing readings, Timed to log data at specific intervals and Single-Shot to hold a reading after a specific wait time is reached
- Audible alarms automatically notify you when a set offset value, high/low limit value and calibration time interval are reached
- Easily access, update and switch your preferred measurement settings, using 10 password-protected methods per channel
- Uniformly mix solutions using up to two meter controlled stirrer probes that are easy to position and can be quickly rinsed between samples
- Data log collects and date/time stamps up to 2000 measurement sets with optional electrode ID, sample ID and account ID
- Calibration log stores 30 most recent calibrations per parameter with viewable graph for multipoint pH and ISE calibrations
- Ensure measurement integrity by viewing the active calibration log while taking readings
- Non-volatile meter memory preserves data and settings, even in the event of power loss
- Meter facilitates data transfers to a computer via USB or RS232 connections
- IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
- Meters include CE, TUV 3-in-1 and FCC Class A certifications
- 3-year replacement warranty
- Measure pH, ion concentration, mV, relative mV or ORP with temperature
- Perform up to a 6 point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
- Perform up to a 6 point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb or none
- Incremental techniques include single known addition, double known addition, single known subtraction, double known subtraction, analate addition and analate subtraction
- Advanced ISE features include non-linear selectable auto-blank, low range stability and electrode ID with calibration verification
- Select the VSTAR40B meter or VSTAR40B2 kit with two pH/ISE modules to measure multiple samples simultaneously
System Access Feature
Orion Versa Star Pro meters offer a system access feature that is designed to help ensure data integrity by preventing undesired access to various meter functions including date/time setting, data log and calibration log. The System Access feature can be easily turned on or off as the needs of your lab change for ultimate flexibility in operation and ease-of-use. When enabled, the System Access feature allows assignment of a system administrator and up to 10 user accounts with customizable access to instrument settings, measurement settings, calibrations and logs. Use the password protection to secure selected meter functions, while leaving other functions accessible for all users. The System Access feature allows you to customize meter access to meet your specific security and data reporting needs.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
USB (2), RS232 | |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.002 | |
Set point alarm, high/low limit alarm, calibration due alarm | |
pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ | |
1 to 6 | |
CE, TUV 3-1, FCC Class A | |
2000 with date and time stamp | |
Color graphic LCD with user-customizable options | |
pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature | |
-2.000 to 20.000 | |
0.1/0.01/0.001 | |
7.000pH (adjustable), 1.000 ISE (adjustable) | |
English, Spanish, French, Italian, German, Chinese, Portuguese and Korean | |
Non-volatile memory preserves data log, calibration log and meter settings | |
DC input: 9VDC, 1A | |
0.0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb or none | |
±2000.0 mV | |
0.1mV | |
±0.1°C | |
5°C to 45°C (ambient) | |
22.0°F to 221.0° F | |
Manual or automatic with ATC temperature probe | |
28cm x 24cm x 10cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2cm (0.8 in.) L & H | |
1.2 kg, modules add 0.2 kg each |
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR) | |
±0.2 mV or ±0.05% of reading whichever is greater | |
Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit | |
Yes | |
1 point relative mV (RmV) mode, 1 point ORP mode | |
1 to 6 | |
Transfer single, range or all data points to printer or computer | |
4 - customizable with modules | |
IP 54 | |
±2000.0 mV | |
0.0001 minimum, 1 to 4 significant digits (user selectable) | |
-5.0°C to +105.0°C | |
Numeric with menu-specific function keys and dual purpose scroll/shortcut keys | |
Automatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with | |
10 per channel | |
Universal AC adapter (included) | |
±2000.0 mV | |
5 to 85 %, non-condensing | |
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting | |
41°F to 113°F (ambient) | |
1 point temperature offset calibration | |
0.1°C, 0.1°F | |
3 year meter and module warranty | |
2.5 lb., modules add 0.5 lb. each |