missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Versa Star pH / LogR Measurement Module

Add pH, mV, ORP and LogR temperature measurement capabilities to your Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / LogR Measurement Module

Brand:  Thermo Scientific™ VSTAR-LR

Product Code. 11611519

  • £758.59 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Add pH, mV, ORP and LogR temperature measurement capabilities to your Thermo Scientific™ Orion™ Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / LogR Measurement Module. Each Orion™ Versa Star meter has four channels that can be customized with interchangeable measurement modules. The Orion™ Versa Star pH/LogR module offers LogR temperature technology that uses the glass-bulb of most pH electrodes to measure both pH and temperature at the same location – ideal for small sample volumes and samples that are difficult to clean.

Select from five Thermo Scientific Orion Versa Star measurement modules to customize the measurement capabilities of your Orion Versa Star meter. Measurement modules are interchangeable and can be added or removed from each Versa Star meter’s four channels at any time.

Includes Orion Versa Star pH/LogR Measurement Module:

  • pH, mV, relative mV or ORP with temperature using LogR technology, ATC probe or manual input
  • LogR technology uses the glass-bulb of most pH electrodes to measure both pH and temperature at the same location – ideal for small sample volumes and samples that are difficult to clean
  • Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading
  • Module offers a 3-year replacement warranty

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.002 pH
±0.1°C
pH with calibration editing option, relative mV (RmV), ORP, LogR temperature and temperature
Orion Versa Star pH/LogR measurement module
12 x 6 x 9.5 cm (4.7 x 2.4 x 3.8 in.)
Versa Star pH/LogR measurement module for use with Versa Star Pro meters to measure pH, mV, relative mV, ORP and temperature using LogR technology, ATC probe or manual entry
IP-54
10 per channel with password protection
-2.000 to 20.000
±2000.0 mV
5 to 85%, non-condensing
0.1, 0.01, 0.001
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
41°F to 113°F
1 point temperature offset calibration
Manual or automatic with ATC temperature probe, automatic with pH electrode using LogR mode
3 year module warranty
±0.2 mV or ±0.05% of reading whichever is greater
Set point alarm, high/low limit alarm, calibration due alarm
1 to 6
pH/LogR Measurement Module
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
7.000pH (adjustable)
pH with LogR temperature, mV, relative mV (RmV) or ORP with temperature
Orion Versa Star pH/LogR/mV/RmV/ORP/Temperature Measurement Module
pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe (purchase separately)
±2000.0mV
0.1mV
0.1°F/°C
22.0°F to 221.0°F
-5°C to 105deg;C
5°C to 45°C
0.5 lb.
0.2 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.