missing translation for 'onlineSavingsMsg'
Learn More

ORM2, Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse Polyclonal Antibody

Brand:  Abnova H00005005-B01P.50ug

Product Code. 16182235

  • £280.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Sequence: MALSWVLTVLSLLPLLEAQIPLCANLVPVPITNATLDRITGKWFYIASAFRNEEYNKSVQEIQATFFYFTPNKTEDTIFLREYQTRQNQCFYNSSYLNVQRENGTVSRYEGGREHVAHLLFLRDTKTLMFGSYLDDEKNWGLSFYADKPETTKEQLGEFYEALDCLCIPRSDVMYTDWKKDKCEPLEKQHEKERKQEEGES
Specifications

Specifications

ORM2
Polyclonal
In 1x PBS, pH 7.4
NM_000608
ORM2
ORM2 (NP_000599.1, 1 a.a. ∼ 201 a.a) full-length human protein.
RUO
5005
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Purified
Western Blot
Unconjugated
orosomucoid 2
AGP-B/AGP-B'/AGP2
Mouse
50 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers