missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orthophosphoric acid ≥85%, BAKER ANALYZED™ ACS, J.T. Baker™
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Color | Max. 10 (APHA) |
Melting Point | 28°C |
Density | 1.71 - 1.87 g/cm³ (25°C) |
Heavy Metals (as Pb) | max. 0.001% |
Packaging | Plastic bottle |
Reducing Substances | Passes Test |
Assay Percent Range | min. 85.0% |
Quantity | 1 L |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'