missing translation for 'onlineSavingsMsg'
Learn More

Oseltamivir phosphate, 98%, Thermo Scientific Chemicals

Product Code. 15608725
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204255-11-8
Molecular Formula C16H28N2O4·H3PO4
Molecular Weight (g/mol) 410.4
InChI Key PGZUMBJQJWIWGJ-ONAKXNSWSA-N
Synonym oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1
PubChem CID 78000
ChEBI CHEBI:7799
IUPAC Name ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid
SMILES CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O

Specifications

Color White to Off-White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 410.4
Percent Purity 98%
Chemical Name or Material Oseltamivir phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.