missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSGIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | OSGIN1 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16834344
|
Proteintech
15248-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16824344
|
Proteintech
15248-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
OSGIN1 regulates the differentiation and proliferation of normal cells through the regulation of cell death.Specifications
OSGIN1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q9UJX0 | |
171493, 29948, 71839 | |
OSGIN1 Fusion Protein Ag4295 | |
Primary | |
-20°C | |
OSGIN1 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BDGI, huOKL38, OKL38, OSGIN1 | |
Osgin1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |