missing translation for 'onlineSavingsMsg'
Learn More
Osmium 99.9%, Thermo Scientific Chemicals Product Code.: 10531844

Osmium 99.9%, Thermo Scientific Chemicals

Product Code. 10531844
1 g, Glass bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10531844

Brand: Thermo Scientific Acros 385160010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-04-2
Molecular Formula Os
Molecular Weight (g/mol) 190.23
MDL Number MFCD00011147
InChI Key SYQBFIAQOQZEGI-UHFFFAOYSA-N
Synonym ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder
PubChem CID 23937
ChEBI CHEBI:30687
IUPAC Name osmium
SMILES [Os]

Specifications

Color Black
Melting Point 3045.0°C
Boiling Point 5027.0°C
Assay Percent Range 99.90%
Linear Formula Os
Quantity 1 g
Merck Index 15, 6988
Solubility Information Solubility in water: insoluble.
Formula Weight 190.2
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Form Powder
Chemical Name or Material Osmium
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435898 Certificate TypeCertificate of Analysis Date03/05/2024 Catalog Number385160050,385160010
Lot NumberA0425005 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number385160050,385160010
Lot NumberA0432933 Certificate TypeCertificate of Analysis Date09/08/2021 Catalog Number385160050,385160010
Lot NumberA0431315 Certificate TypeCertificate of Analysis Date15/06/2021 Catalog Number385160050,385160010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Osmium 99.9%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.