missing translation for 'onlineSavingsMsg'
Learn More

Osmium(III) chloride hydrate

Product Code. 10721555 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
This item is not returnable. View return policy

Product Code. 10721555

Brand: Thermo Scientific Acros 318700025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14996-60-2 | Cl3H2OOs | 314.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14996-60-2
Molecular Formula Cl3H2OOs
Molecular Weight (g/mol) 314.60
MDL Number MFCD00149815
InChI Key KHHOLOMEVBPRQY-UHFFFAOYSA-K
Synonym osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1
PubChem CID 16211485
SMILES O.Cl[Os](Cl)Cl

Specifications

Melting Point >500.0°C
Color Black to Red
Physical Form Crystals or Powder
Quantity 2.5 g
Linear Formula OsCl3·xH2O
Solubility Information Solubility in water: soluble
IUPAC Name trichloroosmium hydrate
Formula Weight 296.56
Packaging Glass bottle
Chemical Name or Material Osmium(III) chloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.