Learn More
Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 197450050
112.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Osmium tetroxide | |
White to Yellow | |
5 mL | |
OsO4 | |
01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236 | |
15, 6990 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
tetraoxoosmium | |
30318 | |
2.5 wt% Min. | |
0.811 |
20816-12-0, 75-65-0 | |
Crystals, Fused Mass or Solution | |
O4Os | |
MFCD00011150 | |
4°C | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
O=[Os](=O)(=O)=O | |
254.23 | |
254.2 | |
Glass bottle | |
0.8110g/mL |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Osmium tetroxide, 2.5 wt% solution in t-butanol, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H301-Toxic if swallowed.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0458596 | Certificate of Analysis | 04/06/2024 | 197450050,197450250,197450010 |
A0451633 | Certificate of Analysis | 17/04/2024 | 197450050,197450250,197450010 |
A0448639 | Certificate of Analysis | 11/04/2024 | 197450050,197450250,197450010 |
A0406410 | Certificate of Analysis | 13/12/2022 | 197450050,197450250,197450010 |
A0401042 | Certificate of Analysis | 13/12/2022 | 197450050,197450250,197450010 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only