missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagent
Specifications
Specifications
Boiling Point | 130.0°C |
Melting Point | 40.0°C to 42.0°C |
Color | Colorless to Yellow |
Physical Form | Crystals |
Quantity | 100 mg |
Assay Percent Range | 99.9+% |
Linear Formula | OsO4 |
Fieser | 01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236 |
Merck Index | 15, 6990 |
Solubility Information | Passes test |
Show More |
Safety and Handling
Product Identifier
- Osmium(VIII)-tetroxide
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P330-Rinse mouth.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction