missing translation for 'onlineSavingsMsg'
Learn More

Osmium Tetroxide, Honeywell™

Product Code. 15611730
Click to view available options
Quantity:
100 mg
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99.8%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Quantity 1 g
Linear Formula OsO4
UN Number 2471
Formula Weight 254.23g/mol
Percent Purity 99%
Grade ReagentPlus™
Vapor Pressure 7mmHg (20°C)
Density Vapor Density: 8.8 (vs air)
Chemical Name or Material Osmium Tetroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'