missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osmium Tetroxide Solution, Honeywell Fluka™
2.5 wt. % in tert-butanol
£118.73 - £351.97
Chemical Identifiers
CAS | 20816-12-0 |
---|---|
Molecular Formula | O4Os |
Molecular Weight (g/mol) | 254.23 |
MDL Number | MFCD00011150 |
InChI Key | VUVGYHUDAICLFK-UHFFFAOYSA-N |
Synonym | osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 |
PubChem CID | 30318 |
IUPAC Name | tetraoxoosmium |
SMILES | O=[Os](=O)(=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
20816-12-0 | |
254.23 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
30318 | |
O=[Os](=O)(=O)=O |
O4Os | |
MFCD00011150 | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
tetraoxoosmium |
Specifications
20816-12-0 | |
O4Os | |
MFCD00011150 | |
4°C (39.2°F) | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
tetraoxoosmium | |
30318 | |
6.76psi (55°C), 5.42psi (20°C) | |
Osmium Tetroxide Solution, Contains tert-butyl hydroperoxide as stabilizer |
Liquid | |
OsO4 | |
UN3286 | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
O=[Os](=O)(=O)=O | |
254.23 | |
254.23g/mol | |
1.0g/mL at 20°C, 0.811g/mL at 25°C |
Safety and Handling
P210-P280-P304 + P340 + P310-P305 + P351 + P338 + P310-P370 + P378-P403 + P235
H225-H301-H312-H315-H317-H318-H330-H334-H335-H336-H341
missing translation for 'dotInformation' : 8(6.1)(3)/PGII
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Osmium Tetroxide Solution, Honeywell Fluka™