missing translation for 'onlineSavingsMsg'
Learn More
Osmium(VIII) oxide, 99.8% (metals basis), Os 74.4% min, Thermo Scientific Chemicals Product Code.: 11346439

Osmium(VIII) oxide, 99.8% (metals basis), Os 74.4% min, Thermo Scientific Chemicals

Product Code. 11346439
0.5 g
Click to view available options
Quantity:
0.5 g
1 g
10 x 1 g
This item is not returnable. View return policy

Product Code. 11346439

Brand: Thermo Scientific Alfa Aesar 012103.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Oxidation, catalyst in Sharpless dihydroxylations Osmium(VIII) oxide is used in the Sharpless oxyamination to give vicinal amino-alcohols. It is used to oxidize alkenes to the vicinal diols. It acts as a staining agent used in transmission electron microscopy to provide contrast to the image and as a stain for lipids in optical microscopy. As a polymer stain, it is also used to stain selectively copolymers especially styrene-butadiene block copolymers. Further, it is involved in the synthesis of Isosteviol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Boiling Point 130°C
Melting Point 40.6°C
Physical Form Crystalline
Quantity 0.5 g
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in chloroform,alcohol and ethers.Soluble in water,organic solvents,benzene,alcohol,ether,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride.
Formula Weight 254.1
Odor Like chlorine
Appearance Crystalline
Vapor Pressure 10 hPa (8mm Hg) at 20°C
Chemical Name or Material Osmium(VIII) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Osmium(VIII) oxide, 99.8% (metals basis), Os 74.4% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.