missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osmolality Standard 500 mOsm/Kg, Reagecon

Used to calibrate, control or validate all types of osmometers.
Brand: Reagecon RE-OSM-500
This item is not returnable.
View return policy
Description
- Osmolality Reference Standard, Osmolality Reference Material.
- The Osmolality results are reported in milliOsmoles per kilogram water (mOsm/kg H2O shortened to mOsm/kg for routine use).
- This product is manufactured in compliance with European, United States and Chinese Pharmacopoeia guidelines where appropriate.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref 264T).
- Prepared gravimetrically on a weight/weight basis.
- Both solute and solvent are weighed on a balance calibrated by Reagecon engineers using OIML traceable weights.
- Reagecon holds ISO 17025 accreditation for calibration of laboratory balances (INAB Ref:265C).
- The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
- The certified values of each standard are verified using a high-performance osmometer calibrated with high quality reference materials.
Specifications
Amber Glass Ampule | |
Liquid | |
Calibration of Osmometer |
12 x 5 mL | |
500 mOsm/Kg |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
OSM50024G1 | Certificate of Analysis | 26/03/2025 | RE-OSM-500 |
OSM50023K1 | Certificate of Analysis | 18/07/2024 | RE-OSM-500 |
OSM50022L1 | Certificate of Analysis | 05/09/2023 | RE-OSM-500 |
OSM50022B1 | Certificate of Analysis | 13/09/2022 | RE-OSM-500 |
OSM50021F1 | Certificate of Analysis | 23/11/2021 | RE-OSM-500 |
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Osmolality Standard 500 mOsm/Kg, Reagecon > 12 x 5 mL
Spot an opportunity for improvement?Share a Content Correction