missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ouchterlony Plates

Product Code. 11423966 Shop All MP Biomedicals Products
Click to view available options
For Use With (Application):
Specific and low affinity antisera
Type:
Ouchterlony Plate
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 11423966

Brand: MP Biomedicals™ 08642771

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for specific but low affinity antisera that requires properly spaced wells and olyethylene glycol in the agarose for optimal immuno -precipitation.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10 μL
Material Polystyrene
Type Ouchterlony Plate
Well Count 7
For Use With (Application) Specific and low affinity antisera
Includes Agarose 1.2% and PEG 5%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.