missing translation for 'onlineSavingsMsg'
Learn More

OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen 17-5743-80

 View more versions of this product

Product Code. 15370740

  • £89.75 / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Description: The 25-D1.16 monoclonal antibody reacts with the ovalbumin-derived peptide SIINFEKL bound to H-2Kb of MHC class I, but not with unbound H-2Kb, or H-2Kb bound with an irrelevant peptide. This antibody has proven to be very useful tracking the quantity and localization of these specific antigen-presenting cells (APC) in vivo. Applications Reported: This eBio25-D1.16 (25-D1.16) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio25-D1.16 (25-D1.16) antibody has been tested by flow cytometric analysis of SIINFEKL peptide pulsed mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Cells can be pulsed with the SIINFEKL peptide according to the following protocol: 1. With cells in flow staining buffer, add SIINFEKL peptide to a final concentration of 30 uM. 2. Incubate cells at 37°C for 2 hours. 3. Wash cells with flow staining buffer. 4. Proceed with cell surface staining as normal. For additional information see the references listed below. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal antibody specifically detects OVA257-264 (SIINFEKL) peptide bound to H-2Kb in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OVA257-264 (SIINFEKL) peptide bound to H-2Kb
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Mouse
25 μg
Primary
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
eBio25-D1.16 (25-D1.16)
APC
H-2Kb-SIINFEKL; OVA257-264; OVA257-264 H-2Kb; OVA-Kb
Affinity chromatography
RUO
Mouse
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only.