Learn More
Fisherbrand™ Oval PTFE Stir Bar
Provides stable stirring action. Fisherbrand; Oval PTFE Stir Bar povides optimal turbulence for round-bottom flasks and containers.
Brand:  Fisherbrand™ 001.650.17
 View more versions of this product
Product Code. 11868952
  • £46.00 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Stable, general-purpose magnetic stir bar
  • Provides excellent turbulence for round-bottom flasks and containers
  • Autoclavable
Specifications
Specifications
Autoclavable
White
Round bottom flasks
17 mm
2/Pk.
PTFE Stirrer Bars Oval
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
PTFE
50 mm
Oval