missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Oval PTFE Stir Bar
Provides stable stirring action. Fisherbrand™ Oval PTFE Stir Bar provides optimal turbulence for round-bottom flasks and containers.
Brand: Fisherbrand™ 001.640.13
Description
- Stable, general-purpose magnetic stir bar
- Provides excellent turbulence for round-bottom flasks and containers
- Autoclavable
Specifications
Autoclavable | |
White | |
Round bottom flasks | |
13 mm | |
5/Pk. | |
PTFE Stirrer Bars Oval |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Alnico V magnet core | |
PTFE | |
40 mm | |
Oval |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Oval PTFE Stir Bar >
Spot an opportunity for improvement?Share a Content Correction