Learn More
Cowie Technology™ Oval PTFE Turbo Stir Bars with Rare Earth Core
Cowie Technology™ Oval PTFE Turbo Stir Bars with Rare Earth Core use rare earth magnets to provide significantly increased magnetic strength compared to Alnico magnets.
Brand:  Cowie Technology™ 001.635.RE
Product Code. 10530120
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Almost totally resistant to demagnetisation
  • Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals