Learn More
Novus Biologicals™ Ovary Tissue Slides (Endometrioid Adenocarcinoma)- Paraffin
Brand: Novus Biologicals™ NBP2-30292
Additional Details : Weight : 0.00970kg
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.Specifications
Human | |
This product is for research use only and is not approved for use in humans or in clinical diagnosis. | |
Endometrioid Adenocarcinoma |
Ovary Tissue Slides (Endometrioid Adenocarcinoma) | |
Human |