Learn More
Labcon Overfit Caps for Culture Tubes
Secure, liquid-tight seal for plastic and glass culture tubes. Available in sizes that fit 10 mm, 12 mm, 13 mm, and 16 mm round bottom culture tubes.
Brand:  Labcon 3403-803-000-9
 View more versions of this product
Product Code. 17244426
  • £17.70 / Pack of 1000
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to easily recap blood drawing tubes, glass test tubes, and plastic culture tubes, Overfit Caps are ideal for protocols that require centrifugation, agitation,
    and refrigeration.
  • Offered in a variety of bright colors, Labcon's Overfit Caps are the ideal alternative to costly parafilm or plug caps that are prone to evaporation.
  • Cap includes integral thumb lever for easy opening and vertical ridges for secure grip
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Made in USA
Specifications
Specifications
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity
No
Not Autoclavable
Orange
1000/Bag, 10 Bags/Cs.
Non-sterile
Single-use
13 mm
Overfit Cap
Polyethylene USP Class VI
Protocols that require centrifugation, agitation, and refrigeration
ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
Culture Tubes