Learn More
Aqualytic™ Overflow Measuring Flask
Aqualytic™ Overflow Measuring Flask is a suitable accessory for BD 600 and BD 606 respirometric method oxygen consumption measuring systems.
Brand:  Aqualytic™ 418660
Product Code. 10079711
  • £42.30 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Suitable for BD 600 or BD 606 respirometric biochemical oxygen demand measuring system
  • Large volume flask
Specifications
Specifications
Overflow Flask
428 mL